Gene Transcripts

Gene Symbol
Sec14l5
Entrez Gene ID
665119
Gene Name
SEC14-like 5 (S. cerevisiae)
Gene Aliases
Chromosome Location
Chr. 16: 5146901 - 5187271 on Build GRCm38
UniGene ID
Mm.102072
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127725.1 NP_001121197.1 8 952
GenBank mRNA BC157907.1 - 8 1018
BC157990.1 - 8 952 9007304

View Full Product Details