Gene Transcripts

Gene Symbol
Rwdd3
Entrez Gene ID
66568
Gene Name
RWD domain containing 3
Gene Aliases
2510027J23Rik, 3110037C01Rik, AI035684, AI662458, Rsume, X2CR1
Chromosome Location
Chr. 3: 121155400 - 121171780 on Build GRCm38
UniGene ID
Mm.263163
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK011007.1 - 3 337
AK011008.1 - 3 337
AK050254.1 - 2 204
BC150838.1 - 3 337 9087749
BY710389.1 - 3 337

View Full Product Details