Gene Transcripts

Gene Symbol
MTX2
Entrez Gene ID
10651
Gene Name
metaxin 2
Gene Aliases
Chromosome Location
Chr. 2: 176269395 - 176338025 on Build GRCh38
UniGene ID
Hs.470728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001006635.2 NP_001006636.1 5, 6 650
NM_001319097.1 NP_001306026.1 4, 5 446
NM_001319098.1 NP_001306027.1 4, 5 446
NM_006554.4 NP_006545.1 4, 5 446
XM_011510497.2 XP_011508799.1 5, 6 543
GenBank mRNA AF053551.1 - 4, 5 419
AK001287.1 - 4, 5 398
AK289359.1 - 4, 5 397
AK303564.1 - 5, 6 575
AK315918.1 - 5, 6 586
BC017271.2 - 5, 6 580 4635884
BC067831.1 - 4, 5 425 5298149
BC088359.1 - 4, 5 381 30378040
BG699784.1 - 5, 6 654
BU165147.1 - 4, 5 377
CX165048.1 - 4, 5 339

View Full Product Details