Gene Transcripts

Gene Symbol
Gp6
Entrez Gene ID
243816
Gene Name
glycoprotein 6 (platelet)
Gene Aliases
9830166G18Rik, Gm469, Gpvi
Chromosome Location
Chr. 7: 4362853 - 4397905 on Build GRCm38
UniGene ID
Mm.248662
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163014.1 NP_001156486.1 6 755
GenBank mRNA BC138170.1 - 6 739

View Full Product Details