Gene Transcripts

Gene Symbol
Mmp17
Entrez Gene ID
288626
Gene Name
matrix metallopeptidase 17
Gene Aliases
Chromosome Location
Chr. 12: 30353863 - 30356836 on Build Rnor_6.0
UniGene ID
Rn.7971
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001105925.1 NP_001099395.1 10 1639
XM_008769195.1 XP_008767417.1 10 2922

View Full Product Details