Gene Transcripts

Gene Symbol
Mesp2
Entrez Gene ID
293046
Gene Name
mesoderm posterior basic helix-loop-helix transcription factor 2
Gene Aliases
Chromosome Location
Chr. 1: 141550633 - 141553239 on Build Rnor_6.0
UniGene ID
Rn.217791
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106273.1 NP_001099743.1 1 789

View Full Product Details