Gene Transcripts

Gene Symbol
Zbtb6
Entrez Gene ID
366029
Gene Name
zinc finger and BTB domain containing 6
Gene Aliases
Zfp482, Znf482
Chromosome Location
Chr. 3: 21679181 - 21697447 on Build Rnor_6.0
UniGene ID
Rn.39069
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108953.1 NP_001102423.1 2 736
XM_006234101.2 XP_006234163.1 2 832
XM_006234102.1 XP_006234164.1 2 1107

View Full Product Details