Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_031439.3 | NP_113627.1 | 2 | 1872 | |
GenBank mRNA | AK055556.1 | - | 2 | 1810 | |
BC004299.1 | - | 2 | 1799 | ||
BC071947.1 | - | 2 | 1798 | 6459570 | |
BU187815.1 | - | 1 | 191 |