Gene Transcripts

Gene Symbol
WDR66
Entrez Gene ID
144406
Gene Name
WD repeat domain 66
Gene Aliases
CaM-IP4
Chromosome Location
Chr. 12: 121918557 - 122003927 on Build GRCh38
UniGene ID
Hs.709837
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001178003.1 NP_001171474.1 16, 17 2780
NM_144668.5 NP_653269.3 16, 17 2767
GenBank mRNA BC036233.1 - 16, 17 2760

View Full Product Details