Gene Transcripts

Gene Symbol
KRTAP8-1
Entrez Gene ID
337879
Gene Name
keratin associated protein 8-1
Gene Aliases
KAP8.1
Chromosome Location
Chr. 21: 30812697 - 30813252 on Build GRCh38
UniGene ID
Hs.407655
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175857.3 NP_787053.1 1 342
GenBank mRNA AJ457064.1 - 1 342

View Full Product Details