Gene Transcripts

Gene Symbol
ZP1
Entrez Gene ID
22917
Gene Name
zona pellucida glycoprotein 1
Gene Aliases
HEL163, OOMD, OOMD1
Chromosome Location
Chr. 11: 60867542 - 60875693 on Build GRCh38
UniGene ID
Hs.172130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_207341.3 NP_997224.2 2 303
XM_011544853.2 XP_011543155.1 1 46
GenBank mRNA EU794684.1 - 2 303

View Full Product Details