Gene Transcripts

Gene Symbol
EFR3B
Entrez Gene ID
22979
Gene Name
EFR3 homolog B
Gene Aliases
KIAA0953
Chromosome Location
Chr. 2: 25042104 - 25159137 on Build GRCh38
UniGene ID
Hs.4892
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001319099.1 NP_001306028.1 18, 19 2251
NM_014971.1 NP_055786.1 18, 19 2263
XM_011532701.2 XP_011531003.1 18, 19 2236
GenBank mRNA AB023170.1 - 18, 19 2237
AK095190.1 - 18, 19 2251
BC049384.1 - 15, 16 1807 5742159

View Full Product Details