Gene Transcripts
- Gene Symbol
- CIAO1
- Entrez Gene ID
- 9391
- Gene Name
- cytosolic iron-sulfur assembly component 1
- Gene Aliases
- CIA1, WDR39
- Chromosome Location
- Chr. 2: 96266146 - 96274179 on Build GRCh38
- UniGene ID
- Hs.12109
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_004804.2 |
NP_004795.1
|
6, 7 |
985 |
|
GenBank mRNA |
AK056423.1 |
- |
4, 5 |
1351 |
|
AK223257.1 |
- |
6, 7 |
906 |
|
AK299503.1 |
- |
6, 7 |
672 |
|
AK315568.1 |
- |
6, 7 |
894 |
|
BC001395.2 |
- |
6, 7 |
849 |
3050587 |
BC032812.1 |
- |
6, 7 |
879 |
5219740 |
CR456802.1 |
- |
6, 7 |
780 |
|
EF011618.1 |
- |
6, 7 |
879 |
|
U63810.1 |
- |
6, 7 |
936 |
|
View Full Product Details