Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001098833.1 | NP_001092303.1 | 11 | 861 | |
NM_020218.1 | NP_064603.1 | 11 | 882 | ||
XM_011525038.2 | XP_011523340.1 | 11 | 1533 | ||
XM_011525044.2 | XP_011523346.1 | 11 | 1476 | ||
XM_011525046.2 | XP_011523348.1 | 11 | 1455 | ||
XM_017024880.1 | XP_016880369.1 | 12 | 1111 | ||
XM_017024881.1 | XP_016880370.1 | 12 | 1410 | ||
XM_017024882.1 | XP_016880371.1 | 11 | 1512 | ||
XM_017024883.1 | XP_016880372.1 | 12 | 1114 | ||
XM_017024884.1 | XP_016880373.1 | 11 | 1136 | ||
XM_017024885.1 | XP_016880374.1 | 12 | 1057 | ||
XM_017024886.1 | XP_016880375.1 | 12 | 1036 | ||
GenBank mRNA | AK056002.1 | - | 8 | 405 | |
AK126117.1 | - | 11 | 1079 | ||
AK296689.1 | - | 5 | 294 | ||
AL390158.1 | - | 6 | 291 | ||
BC024695.2 | - | 4 | 219 | ||
BC037418.1 | - | 11 | 789 | ||
BC113595.1 | - | 11 | 861 | 8322647 | |
BC126113.1 | - | 11 | 861 | 8991829 |