Gene Transcripts

Gene Symbol
Ctxn3
Entrez Gene ID
629147
Gene Name
cortexin 3
Gene Aliases
ENSMUSG00000069372
Chromosome Location
Chr. 18: 57467656 - 57478134 on Build GRCm38
UniGene ID
Mm.57415
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134697.1 NP_001128169.1 2 501
GenBank mRNA AK042789.1 - 2 461

View Full Product Details