Gene Transcripts

Gene Symbol
Kcnk15
Entrez Gene ID
241769
Gene Name
potassium channel, subfamily K, member 15
Gene Aliases
KCNK11, KCNK14, KT3.3, TASK5
Chromosome Location
Chr. 2: 163853750 - 163858874 on Build GRCm38
UniGene ID
Mm.295337
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001030292.1 NP_001025463.1 2 449
GenBank mRNA BC147255.1 - 2 548
BC147256.1 - 2 548 9056288

View Full Product Details