Gene Transcripts

Gene Symbol
Bean1
Entrez Gene ID
65115
Gene Name
brain expressed, associated with Nedd4, 1
Gene Aliases
Bean
Chromosome Location
Chr. 8: 104170376 - 104219098 on Build GRCm38
UniGene ID
Mm.41849
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001141924.1 NP_001135396.1 3, 4 845
GenBank mRNA AK134633.1 - 3, 4 845

View Full Product Details