Gene Transcripts

Gene Symbol
Vmn2r2
Entrez Gene ID
100125586
Gene Name
vomeronasal 2, receptor 2
Gene Aliases
ENSMUSG00000043897
Chromosome Location
Chr. 3: 64115730 - 64140657 on Build GRCm38
UniGene ID
Mm.469888
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001104592.1 NP_001098062.1 2 548

View Full Product Details