Gene Transcripts

Gene Symbol
Ercc6
Entrez Gene ID
319955
Gene Name
excision repair cross-complementing rodent repair deficiency, complementation group 6
Gene Aliases
4732403I04, C130058G22Rik, CSB
Chromosome Location
Chr. 14: 32513288 - 32580990 on Build GRCm38
UniGene ID
Mm.318310
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081221.1 NP_001074690.1 9 2004
GenBank mRNA BC132447.1 - 5 943

View Full Product Details