Gene Transcripts

Gene Symbol
Dpm3
Entrez Gene ID
502017
Gene Name
dolichyl-phosphate mannosyltransferase polypeptide 3
Gene Aliases
RGD1561807
Chromosome Location
Chr. 2: 188583664 - 188584179 on Build Rnor_6.0
UniGene ID
Rn.20619
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109331.2 NP_001102801.1 2 228

Gene Symbol
LOC100910177
Entrez Gene ID
100910177
Gene Name
dolichol-phosphate mannosyltransferase subunit 3-like
Gene Aliases
Chromosome Location
Chr. 15: 52919250 - 52919657 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_003751511.3 XP_003751559.2 1 217

View Full Product Details