Gene Transcripts

Gene Symbol
Rhbdl2
Entrez Gene ID
298512
Gene Name
rhomboid, veinlet-like 2 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 5: 141462300 - 141512067 on Build Rnor_6.0
UniGene ID
Rn.227169
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106684.1 NP_001100154.1 3 324
XM_006238802.2 XP_006238864.1 5 932

View Full Product Details