Gene Transcripts

Gene Symbol
Wdr35l
Entrez Gene ID
503018
Gene Name
WD repeat domain 35-like
Gene Aliases
RGD1564116, Wdr35
Chromosome Location
Chr. 6: 34094290 - 34152048 on Build Rnor_6.0
UniGene ID
Rn.14574
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099340.1 NP_001092810.1 20, 21 2526
XM_006239885.2 XP_006239947.1 21, 22 2575
GenBank mRNA EF613262.1 - 20, 21 2526

View Full Product Details