Gene Transcripts

Gene Symbol
Phactr1
Entrez Gene ID
306844
Gene Name
phosphatase and actin regulator 1
Gene Aliases
Chromosome Location
Chr. 17: 23245423 - 23761207 on Build Rnor_6.0
UniGene ID
Rn.22981
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_214457.2 NP_999622.2 13 1869
GenBank mRNA AY494977.1 - 1738
BC098634.1 - 13 1869 7317963

View Full Product Details