Gene Transcripts

Gene Symbol
Spag16
Entrez Gene ID
501158
Gene Name
sperm associated antigen 16
Gene Aliases
RGD1565062
Chromosome Location
Chr. 9: 77027626 - 77833476 on Build Rnor_6.0
UniGene ID
Rn.48315
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134728.1 NP_001128200.1 8 1404
GenBank mRNA BC158602.1 - 8 1404

View Full Product Details