Gene Transcripts

Gene Symbol
SPEF1
Entrez Gene ID
25876
Gene Name
sperm flagellar 1
Gene Aliases
C20orf28, CLAMP, SPEF1A
Chromosome Location
Chr. 20: 3777504 - 3781466 on Build GRCh38
UniGene ID
Hs.72620
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_015417.4 NP_056232.2 7 1063
XM_005260683.4 XP_005260740.1 7 1022
GenBank mRNA AL080154.1 - 6 1189
BC022476.1 - 5 1367 4816764

View Full Product Details