Gene Transcripts

Gene Symbol
FBXO10
Entrez Gene ID
26267
Gene Name
F-box protein 10
Gene Aliases
FBX10, PRMT11
Chromosome Location
Chr. 9: 37509920 - 37576459 on Build GRCh38
UniGene ID
Hs.709527
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012166.2 NP_036298.2 9 2498
XM_006716754.3 XP_006716817.1 7 1053
XM_017014618.1 XP_016870107.1 9 2704
XM_017014619.1 XP_016870108.1 9 2815
GenBank mRNA AB209610.1 - 8 2086
AK123986.1 - 1 172
BC125124.1 - 7 1092 40119018
BC125125.1 - 9 2577 40119019
BC140785.1 - 8 2469 9021653
BC171785.1 - 8 2469 9054439

View Full Product Details