Gene Transcripts
- Gene Symbol
- RBMS3
- Entrez Gene ID
- 27303
- Gene Name
- RNA binding motif single stranded interacting protein 3
- Gene Aliases
- Chromosome Location
- Chr. 3: 29280860 - 30010395 on Build GRCh38
- UniGene ID
- Hs.221436
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001003792.2 |
NP_001003792.1
|
8, 9 |
1165 |
|
NM_001003793.2 |
NP_001003793.1
|
8, 9 |
1168 |
|
NM_001177711.1 |
NP_001171182.1
|
8, 9 |
1168 |
|
NM_001177712.1 |
NP_001171183.1
|
9, 10 |
1207 |
|
NM_014483.3 |
NP_055298.2
|
8, 9 |
1168 |
|
XM_005265060.2 |
XP_005265117.1
|
8, 9 |
1617 |
|
XM_005265061.2 |
XP_005265118.1
|
8, 9 |
1620 |
|
XM_005265062.1 |
XP_005265119.1
|
8, 9 |
1620 |
|
XM_005265063.1 |
XP_005265120.1
|
5, 6 |
825 |
|
XM_005265065.4 |
XP_005265122.1
|
5, 6 |
518 |
|
XM_017006178.1 |
XP_016861667.1
|
8, 9 |
1617 |
|
XM_017006179.1 |
XP_016861668.1
|
8, 9 |
1617 |
|
XM_017006180.1 |
XP_016861669.1
|
8, 9 |
1620 |
|
XM_017006181.1 |
XP_016861670.1
|
8, 9 |
1617 |
|
XM_017006182.1 |
XP_016861671.1
|
9, 10 |
1696 |
|
GenBank mRNA |
AF023259.1 |
- |
8, 9 |
945 |
|
AK292789.1 |
- |
8, 9 |
844 |
|
AY236871.1 |
- |
8, 9 |
1028 |
|
AY236872.1 |
- |
8, 9 |
1025 |
|
BC117315.1 |
- |
9, 10 |
904 |
40125866 |
BC143521.1 |
- |
9, 10 |
904 |
9052030 |
View Full Product Details