Gene Transcripts
- Gene Symbol
- TNRC6A
- Entrez Gene ID
- 27327
- Gene Name
- trinucleotide repeat containing 6A
- Gene Aliases
- CAGH26, GW1, GW182, TNRC6
- Chromosome Location
- Chr. 16: 24610260 - 24826227 on Build GRCh38
- UniGene ID
- Hs.655057
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014494.2 |
NP_055309.2
|
17, 18 |
4648 |
|
XM_005255254.3 |
XP_005255311.1
|
16, 17 |
4491 |
|
XM_005255257.4 |
XP_005255314.1
|
16, 17 |
4615 |
|
XM_017023143.1 |
XP_016878632.1
|
16, 17 |
4592 |
|
XM_017023144.1 |
XP_016878633.1
|
15, 16 |
4559 |
|
XM_017023145.1 |
XP_016878634.1
|
16, 17 |
4525 |
|
XM_017023146.1 |
XP_016878635.1
|
16, 17 |
4657 |
|
XM_017023147.1 |
XP_016878636.1
|
16, 17 |
4685 |
|
XM_017023148.1 |
XP_016878637.1
|
15, 16 |
4412 |
|
XM_017023149.1 |
XP_016878638.1
|
14, 15 |
4409 |
|
XM_017023150.1 |
XP_016878639.1
|
15, 16 |
4374 |
|
XM_017023151.1 |
XP_016878640.1
|
14, 15 |
4263 |
|
XM_017023152.1 |
XP_016878641.1
|
15, 16 |
4164 |
|
XM_017023153.1 |
XP_016878642.1
|
16, 17 |
4615 |
|
XM_017023154.1 |
XP_016878643.1
|
14, 15 |
4506 |
|
GenBank mRNA |
AB040893.1 |
- |
12, 13 |
2848 |
|
AJ492221.1 |
- |
16, 17 |
4534 |
|
AK025696.1 |
- |
10, 11 |
1205 |
|
AK094542.1 |
- |
4, 5 |
1230 |
|
AL713750.1 |
- |
4, 5 |
1401 |
|
AL833757.1 |
- |
11, 12 |
1809 |
|
AY035864.1 |
- |
14, 15 |
4182 |
|
View Full Product Details