Gene Transcripts
- Gene Symbol
- GPN3
- Entrez Gene ID
- 51184
- Gene Name
- GPN-loop GTPase 3
- Gene Aliases
- ATPBD1C
- Chromosome Location
- Chr. 12: 110452486 - 110468835 on Build GRCh38
- UniGene ID
- Hs.634680
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC008416.1 |
- |
7 |
783 |
|
View Full Product Details