Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317724.1 | NP_001304653.1 | 6 | 609 | |
NM_016167.4 | NP_057251.2 | 6 | 609 | ||
GenBank mRNA | AF172066.3 | - | 6 | 609 | |
AK075301.1 | - | 6 | 592 | ||
AL050299.1 | - | 5 | 309 | ||
BC023517.2 | - | 6 | 584 | 4298095 | |
BC062683.1 | - | 6 | 605 | 6499123 | |
BU071935.1 | - | 6 | 362 |