Gene Transcripts
- Gene Symbol
- GPR89B
- Entrez Gene ID
- 51463
- Gene Name
- G protein-coupled receptor 89B
- Gene Aliases
- GPHR, GPR89, GPR89C, SH120, UNQ192
- Chromosome Location
- Chr. 1: 147928393 - 147993523 on Build GRCh38
- UniGene ID
- Hs.504320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016334.3 |
NP_057418.1
|
11, 12 |
1148 |
|
XM_005277401.3 |
XP_005277458.1
|
11, 12 |
1121 |
|
XM_005277402.4 |
XP_005277459.1
|
11, 12 |
1285 |
|
XM_005277404.3 |
XP_005277461.1
|
11, 12 |
1141 |
|
XM_006711377.3 |
XP_006711440.1
|
12, 13 |
1184 |
|
XM_006711378.2 |
XP_006711441.1
|
12, 13 |
1187 |
|
XM_011509610.2 |
XP_011507912.1
|
12, 13 |
1181 |
|
XM_011509612.2 |
XP_011507914.1
|
12, 13 |
1142 |
|
XM_017001448.1 |
XP_016856937.1
|
12, 13 |
1684 |
|
GenBank mRNA |
AF151035.1 |
- |
3, 4 |
260 |
|
AK299995.1 |
- |
11, 12 |
1119 |
|
AK302266.1 |
- |
11, 12 |
1140 |
|
AL133020.1 |
- |
11, 12 |
1649 |
|
CD102528.1 |
- |
6, 7 |
513 |
|
KJ534849.1 |
- |
11, 12 |
1010 |
|
KJ534850.1 |
- |
11, 12 |
1004 |
|
KJ534851.1 |
- |
11, 12 |
935 |
|
KJ535070.1 |
- |
12, 13 |
1332 |
|
U78723.1 |
- |
11, 12 |
1107 |
|
- Gene Symbol
- GPR89A
- Entrez Gene ID
- 653519
- Gene Name
- G protein-coupled receptor 89A
- Gene Aliases
- GPHR, GPR89, GPR89B, SH120, UNQ192
- Chromosome Location
- Chr. 1: 145607960 - 145670977 on Build GRCh38
- UniGene ID
- Hs.504320
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001097612.1 |
NP_001091081.1
|
11, 12 |
1178 |
|
NM_001097613.2 |
NP_001091082.2
|
12, 13 |
1224 |
|
NR_036541.1 |
|
11, 12 |
1198 |
|
XM_006711492.3 |
XP_006711555.1
|
11, 12 |
1148 |
|
XM_006711493.3 |
XP_006711556.1
|
12, 13 |
1156 |
|
XM_011509908.2 |
XP_011508210.1
|
12, 13 |
1194 |
|
XM_011509909.2 |
XP_011508211.1
|
11, 12 |
1406 |
|
GenBank mRNA |
AB097024.1 |
- |
11, 12 |
1119 |
|
AB097025.1 |
- |
11, 12 |
1119 |
|
AF132947.1 |
- |
11, 12 |
1103 |
|
AK001354.1 |
- |
4, 5 |
1916 |
|
AK021758.1 |
- |
11, 12 |
1168 |
|
AK094228.1 |
- |
2, 3 |
1119 |
|
AK223223.1 |
- |
11, 12 |
1125 |
|
AK301939.1 |
- |
11, 12 |
1025 |
|
AK316371.1 |
- |
12, 13 |
1136 |
|
AY358966.1 |
- |
11, 12 |
1153 |
|
BC003187.1 |
- |
11, 12 |
1081 |
3537949 |
BC067816.1 |
- |
11, 12 |
1137 |
5295014 |
BC146880.1 |
- |
11, 12 |
1099 |
9056808 |
BT006679.1 |
- |
11, 12 |
1004 |
|
KJ534847.1 |
- |
11, 12 |
1004 |
|
KJ534848.1 |
- |
11, 12 |
1010 |
|
KJ535029.1 |
- |
10, 11 |
1004 |
|
KJ535049.1 |
- |
11, 12 |
1024 |
|
View Full Product Details