Gene Transcripts

Gene Symbol
CDK5RAP1
Entrez Gene ID
51654
Gene Name
CDK5 regulatory subunit associated protein 1
Gene Aliases
C20orf34, C42, CGI-05, HSPC167
Chromosome Location
Chr. 20: 33358839 - 33402768 on Build GRCh38
UniGene ID
Hs.435952
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278167.1 NP_001265096.1 8, 9 1262
NM_001278169.1 NP_001265098.1 8, 9 1030
NM_016082.4 NP_057166.4 8, 9 1114
NM_016408.3 NP_057492.2 8, 9 1262
XM_011528856.2 XP_011527158.1 5, 6 1998
XM_011528857.2 XP_011527159.1 7, 8 874
XM_011528858.1 XP_011527160.1 7, 8 1113
XM_011528859.2 XP_011527161.1 7, 8 873
XM_011528860.1 XP_011527162.1 5, 6 573
XM_017027876.1 XP_016883365.1 7, 8 1012
XM_017027877.1 XP_016883366.1 2, 3 326
GenBank mRNA AF152097.1 - 9, 10 1240
AF161516.1 - 8, 9 1210
AK023992.1 - 8, 9 1212
AK027449.1 - 8, 9 1062
AK222745.1 - 8, 9 1212
AK292075.1 - 8, 9 980
AK295342.1 - 8, 9 1198
AK314532.1 - 8, 9 1212
AY462283.1 - 8, 9 1213
AY462284.1 - 8, 9 1213
BC050706.1 - 8, 9 1205 4418659
CX785345.1 - 3, 4 469
KU178645.1 - 7, 8 1100

View Full Product Details