Gene Transcripts

Gene Symbol
CDR1
Entrez Gene ID
1038
Gene Name
cerebellar degeneration related protein 1
Gene Aliases
CDR, CDR34, CDR62A
Chromosome Location
Chr. X: 140783260 - 140784558 on Build GRCh38
UniGene ID
Hs.446675
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004065.2 NP_004056.2 1 323
GenBank mRNA BC113472.1 - 1 248
BC113474.1 - 1 248 8322526
M16965.1 - 1 191

View Full Product Details