Gene Transcripts
- Gene Symbol
- CLIC5
- Entrez Gene ID
- 53405
- Gene Name
- chloride intracellular channel 5
- Gene Aliases
- DFNB102, DFNB103, MST130, MSTP130
- Chromosome Location
- Chr. 6: 45898451 - 46129809 on Build GRCh38
- UniGene ID
- Hs.485489
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114086.1 |
NP_001107558.1
|
1, 2 |
654 |
|
NM_001256023.1 |
NP_001242952.1
|
1, 2 |
418 |
|
NM_016929.4 |
NP_058625.2
|
1, 2 |
418 |
|
XM_011514692.2 |
XP_011512994.1
|
1, 2 |
654 |
|
XM_011514694.2 |
XP_011512996.1
|
1, 2 |
654 |
|
XM_017010953.1 |
XP_016866442.1
|
2, 3 |
899 |
|
GenBank mRNA |
AF176920.1 |
- |
1, 2 |
217 |
|
AF216941.1 |
- |
1, 2 |
368 |
|
AK075144.1 |
- |
1, 2 |
357 |
|
AK075163.1 |
- |
1, 2 |
269 |
|
AK097048.1 |
- |
1, 2 |
654 |
|
AK222544.1 |
- |
1, 2 |
386 |
|
AK223130.1 |
- |
1, 2 |
389 |
|
AK314416.1 |
- |
1, 2 |
234 |
|
AY032602.1 |
- |
1, 2 |
548 |
|
BC035968.1 |
- |
1, 2 |
386 |
4611102 |
DA843541.1 |
- |
1, 2 |
418 |
|
DQ679794.1 |
- |
1, 2 |
386 |
|
View Full Product Details