Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001278722.1 | NP_001265651.1 | 6 | 950 | |
NM_001278723.1 | NP_001265652.1 | 5 | 1090 | ||
NM_001278724.1 | NP_001265653.1 | 5 | 1024 | ||
NM_014432.3 | NP_055247.3 | 6 | 1269 | ||
XM_006715506.2 | XP_006715569.1 | 5 | 666 | ||
XM_011535904.2 | XP_011534206.1 | 5 | 777 | ||
XM_017010954.1 | XP_016866443.1 | 6 | 1458 | ||
XM_017010955.1 | XP_016866444.1 | 6 | 918 | ||
GenBank mRNA | AF184971.1 | - | 6 | 1053 | |
AJ420490.1 | - | 2 | 244 | ||
AK098312.1 | - | 2 | 859 | ||
AK297121.1 | - | 6 | 912 | ||
AK315293.1 | - | 6 | 1057 | ||
AY358883.1 | - | 5 | 604 | ||
BC113574.1 | - | 6 | 1147 | 8322626 | |
BC113602.1 | - | 6 | 1147 | 8322654 |