Gene Transcripts

Gene Symbol
RNLS
Entrez Gene ID
55328
Gene Name
renalase, FAD dependent amine oxidase
Gene Aliases
C10orf59, RENALASE
Chromosome Location
Chr. 10: 88176458 - 88583860 on Build GRCh38
UniGene ID
Hs.149849
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001031709.2 NP_001026879.2 5, 6 830
NM_018363.3 NP_060833.1 5, 6 830
XM_005269946.1 XP_005270003.1 5, 6 756
XM_005269948.2 XP_005270005.1 3, 4 1116
XM_005269950.3 XP_005270007.1 3, 4 1116
XM_011539924.2 XP_011538226.1 5, 6 756
XM_011539927.2 XP_011538229.1 5, 6 756
XM_017016380.1 XP_016871869.1 5, 6 756
XM_017016381.1 XP_016871870.1 5, 6 756
XM_017016382.1 XP_016871871.1 3, 4 1116
XM_017016385.1 XP_016871874.1 2, 3 1710
GenBank mRNA AK002080.1 - 5, 6 757
AK296262.1 - 3, 4 514
BC005364.1 - 5, 6 718 3828014
BX648154.1 - 2, 3 239

View Full Product Details