Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001283.3 | NP_001274.1 | 2 | 152 | |
GenBank mRNA | AB015319.1 | - | 2 | 69 | |
AK312151.1 | - | 2 | 118 | ||
BC003561.1 | - | 2 | 71 | 3504050 | |
BI597121.1 | - | 2 | 157 | ||
BI598615.1 | - | 2 | 157 | ||
BT006779.1 | - | 1 | 42 |