Gene Transcripts
- Gene Symbol
- CLK2
- Entrez Gene ID
- 1196
- Gene Name
- CDC like kinase 2
- Gene Aliases
- Chromosome Location
- Chr. 1: 155262868 - 155273529 on Build GRCh38
- UniGene ID
- Hs.73986
- Species
- Homo sapiens
- Gene Symbol
- CLK2P1
- Entrez Gene ID
- 1197
- Gene Name
- CDC like kinase 2, pseudogene 1
- Gene Aliases
- CLK2P
- Chromosome Location
- Chr. 7: 23584716 - 23586527 on Build GRCh38
- UniGene ID
- Hs.632635
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002711.3 |
|
1 |
1327 |
|
GenBank mRNA |
BC022025.1 |
- |
1 |
1343 |
|
View Full Product Details