Gene Transcripts

Gene Symbol
UBQLN4
Entrez Gene ID
56893
Gene Name
ubiquilin 4
Gene Aliases
A1U, A1Up, C1orf6, CIP75, UBIN
Chromosome Location
Chr. 1: 156035294 - 156053825 on Build GRCh38
UniGene ID
Hs.283739
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001304342.1 NP_001291271.1 4, 5 815
NM_020131.4 NP_064516.2 4, 5 875
XM_005245348.3 XP_005245405.1 4, 5 780
GenBank mRNA AB209566.1 - 4, 5 748
AF113544.1 - 1, 2 281
AF188240.1 - 4, 5 1029
AK302895.1 - 4, 5 704
AK314413.1 - 4, 5 763
BC018403.2 - 4, 5 763 4214386
BC063841.1 - 4, 5 775 6183942

View Full Product Details