Gene Transcripts

Gene Symbol
LYRM4
Entrez Gene ID
57128
Gene Name
LYR motif containing 4
Gene Aliases
C6orf149, CGI-203, COXPD19, ISD11
Chromosome Location
Chr. 6: 5031029 - 5260950 on Build GRCh38
UniGene ID
Hs.387755
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164840.2 NP_001158312.1 2 320
NM_001164841.2 NP_001158313.1 2 320
NM_001318782.1 NP_001305711.1 2 320
NM_001318783.1 NP_001305712.1 2 320
NM_020408.5 NP_065141.3 2 320
NR_104417.1 2 320
NR_134856.1 2 320
XM_017011083.1 XP_016866572.1 2 314
XM_017011084.1 XP_016866573.1 2 317
GenBank mRNA AF088060.1 - 2 100
AF170070.1 - 2 116
AF285118.1 - 2 116
AK291158.1 - 2 112
AK309458.1 - 1 19
AK309661.1 - 2 122
BC009552.2 - 2 171 3898393
BI226933.1 - 2 294
BI753819.1 - 2 306
BI758258.1 - 2 309
BX353584.2 - 2 99
CK819966.1 - 2 96
DA114665.1 - 2 112
DA222333.1 - 2 112
HY168919.1 - 2 321

View Full Product Details