Gene Transcripts
- Gene Symbol
- DOLPP1
- Entrez Gene ID
- 57171
- Gene Name
- dolichyldiphosphatase 1
- Gene Aliases
- LSFR2
- Chromosome Location
- Chr. 9: 129081104 - 129090438 on Build GRCh38
- UniGene ID
- Hs.21701
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001135917.1 |
NP_001129389.1
|
4, 5 |
397 |
|
NM_020438.4 |
NP_065171.2
|
4, 5 |
397 |
|
NR_072983.1 |
|
3, 4 |
312 |
|
GenBank mRNA |
AK290713.1 |
- |
4, 5 |
397 |
|
AK293627.1 |
- |
4, 5 |
388 |
|
AY189675.1 |
- |
4, 5 |
400 |
|
BC009493.2 |
- |
4, 5 |
367 |
|
BC033686.1 |
- |
4, 5 |
374 |
5195127 |
BC040152.1 |
- |
3, 4 |
282 |
|
DA590931.1 |
- |
4, 5 |
397 |
|
View Full Product Details