Gene Transcripts
- Gene Symbol
- COL4A5
- Entrez Gene ID
- 1287
- Gene Name
- collagen type IV alpha 5 chain
- Gene Aliases
- ASLN, ATS, CA54
- Chromosome Location
- Chr. X: 108439844 - 108697545 on Build GRCh38
- UniGene ID
- Hs.369089
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_000495.4 |
NP_000486.1
|
8, 9 |
755 |
|
NM_033380.2 |
NP_203699.1
|
8, 9 |
755 |
|
XM_011530849.2 |
XP_011529151.2
|
9, 10 |
503 |
|
XM_017029259.1 |
XP_016884748.1
|
9, 10 |
503 |
|
XM_017029260.1 |
XP_016884749.1
|
9, 10 |
503 |
|
XM_017029261.1 |
XP_016884750.1
|
9, 10 |
503 |
|
XM_017029262.1 |
XP_016884751.1
|
9, 10 |
503 |
|
GenBank mRNA |
BC151846.1 |
- |
8, 9 |
717 |
|
KM282618.1 |
- |
8, 9 |
755 |
|
M58526.1 |
- |
5, 6 |
231 |
|
M90464.1 |
- |
8, 9 |
675 |
|
View Full Product Details