Gene Transcripts

Gene Symbol
CELF5
Entrez Gene ID
60680
Gene Name
CUGBP, Elav-like family member 5
Gene Aliases
BRUNOL-5, BRUNOL5, CELF-5
Chromosome Location
Chr. 19: 3224676 - 3297076 on Build GRCh38
UniGene ID
Hs.655747
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172673.1 NP_001166144.1 12 1337
NM_021938.3 NP_068757.2 12 1454
NR_033342.1 12 1251
XM_006722832.1 XP_006722895.1 12 1481
XM_006722833.1 XP_006722896.1 12 1478
XM_006722836.2 XP_006722899.1 11 1122
XM_006722837.3 XP_006722900.1 12 1393
XM_006722838.3 XP_006722901.1 11 1265
XM_011528173.1 XP_011526475.1 11 1094
XM_011528174.2 XP_011526476.1 11 1263
XM_017027111.1 XP_016882600.1 10 1337
GenBank mRNA AB209067.1 - 12 1337
AF248649.1 - 2 233
AF329266.1 - 12 1417
AK294109.1 - 12 1251
AL832550.1 - 10 1000
BC028101.1 - 12 1454 5242909

View Full Product Details