Gene Transcripts
- Gene Symbol
- RBSN
- Entrez Gene ID
- 64145
- Gene Name
- rabenosyn, RAB effector
- Gene Aliases
- Rabenosyn-5, ZFYVE20
- Chromosome Location
- Chr. 3: 15070069 - 15099163 on Build GRCh38
- UniGene ID
- Hs.475565
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001302378.1 |
NP_001289307.1
|
10, 11 |
1473 |
|
NM_022340.3 |
NP_071735.2
|
11, 12 |
1629 |
|
XM_005265384.4 |
XP_005265441.1
|
11, 12 |
1592 |
|
XM_005265385.4 |
XP_005265442.1
|
10, 11 |
1447 |
|
XM_011534001.2 |
XP_011532303.1
|
7, 8 |
969 |
|
XM_017007023.1 |
XP_016862512.1
|
10, 11 |
1465 |
|
XM_017007024.1 |
XP_016862513.1
|
12, 13 |
1775 |
|
XM_017007025.1 |
XP_016862514.1
|
11, 12 |
1634 |
|
XM_017007026.1 |
XP_016862515.1
|
10, 11 |
1469 |
|
XM_017007027.1 |
XP_016862516.1
|
10, 11 |
1378 |
|
XM_017007028.1 |
XP_016862517.1
|
10, 11 |
1368 |
|
GenBank mRNA |
AB209673.1 |
- |
11, 12 |
1589 |
|
AK291288.1 |
- |
10, 11 |
1464 |
|
AL833545.1 |
- |
11, 12 |
1623 |
|
AY009133.1 |
- |
8, 9 |
1006 |
|
BC106940.2 |
- |
8, 9 |
1006 |
40034008 |
View Full Product Details