Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146056.2 | NP_001139528.1 | 6 | 2192 | |
NM_001146083.2 | NP_001139555.1 | 7 | 2271 | ||
NM_024298.4 | NP_077274.3 | 8 | 2398 | ||
XM_011527299.2 | XP_011525601.1 | 8 | 2602 | ||
XM_011527300.2 | XP_011525602.1 | 8 | 1993 | ||
GenBank mRNA | AF211969.1 | - | 2 | 849 | |
AK055908.1 | - | 8 | 2331 | ||
AK122789.1 | - | 9 | 2112 | ||
AK123290.1 | - | 2 | 2845 | ||
AK298689.1 | - | 6 | 1878 | ||
BC002512.2 | - | 7 | 1848 | ||
BC003164.1 | - | 8 | 1929 | 2958347 | |
BC015857.1 | - | 7 | 1835 | 4671596 | |
S82470.1 | - | 6 | 1702 |