Gene Transcripts
- Gene Symbol
- ASB13
- Entrez Gene ID
- 79754
- Gene Name
- ankyrin repeat and SOCS box containing 13
- Gene Aliases
- Chromosome Location
- Chr. 10: 5638857 - 5666595 on Build GRCh38
- UniGene ID
- Hs.445899
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024701.3 |
NP_078977.2
|
5, 6 |
748 |
|
NR_024581.1 |
|
4, 5 |
597 |
|
NR_037164.1 |
|
6, 7 |
829 |
|
GenBank mRNA |
AF403031.1 |
- |
5, 6 |
730 |
|
AK023196.1 |
- |
5, 6 |
731 |
|
AK091935.1 |
- |
4, 5 |
597 |
|
AK292071.1 |
- |
5, 6 |
734 |
|
BC012056.1 |
- |
6, 7 |
806 |
4552643 |
BG436652.1 |
- |
4, 5 |
576 |
|
CR457302.1 |
- |
5, 6 |
704 |
|
View Full Product Details