Gene Transcripts
- Gene Symbol
- DNAJB14
- Entrez Gene ID
- 79982
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B14
- Gene Aliases
- EGNR9427, PRO34683
- Chromosome Location
- Chr. 4: 99896248 - 99946726 on Build GRCh38
- UniGene ID
- Hs.577426
- Species
- Homo sapiens
- Gene Symbol
- LOC646358
- Entrez Gene ID
- 646358
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B14 pseudogene
- Gene Aliases
- Chromosome Location
- Chr. 15: 65770250 - 65772154 on Build GRCh38
- UniGene ID
- Hs.669711
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC107769.1 |
- |
1 |
479 |
|
View Full Product Details