Gene Transcripts

Gene Symbol
OR6K2
Entrez Gene ID
81448
Gene Name
olfactory receptor family 6 subfamily K member 2
Gene Aliases
OR1-17
Chromosome Location
Chr. 1: 158699678 - 158700652 on Build GRCh38
UniGene ID
Hs.554489
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001005279.1 NP_001005279.1 1 517
GenBank mRNA BC137022.1 - 1 561
BC137023.1 - 1 602 9021016

View Full Product Details