Gene Transcripts

Gene Symbol
KRTAP2-1
Entrez Gene ID
81872
Gene Name
keratin associated protein 2-1
Gene Aliases
KAP2.1A, KRTAP2.1A
Chromosome Location
Chr. 17: 41046541 - 41047316 on Build GRCh38
UniGene ID
Hs.528921
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001123387.1 NP_001116859.1 1 637
GenBank mRNA AJ296345.1 - 1 493

View Full Product Details