Gene Transcripts
- Gene Symbol
- TMEM263
- Entrez Gene ID
- 90488
- Gene Name
- transmembrane protein 263
- Gene Aliases
- C12orf23
- Chromosome Location
- Chr. 12: 106955535 - 106974037 on Build GRCh38
- UniGene ID
- Hs.257664
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001319661.1 |
NP_001306590.1
|
2, 3 |
420 |
|
NM_001319662.1 |
NP_001306591.1
|
2, 3 |
441 |
|
NM_001319663.1 |
NP_001306592.1
|
4, 5 |
586 |
|
NM_001319664.1 |
NP_001306593.1
|
4, 5 |
604 |
|
NM_152261.3 |
NP_689474.1
|
3, 4 |
488 |
|
XM_017020198.1 |
XP_016875687.1
|
3, 4 |
675 |
|
GenBank mRNA |
AK001731.1 |
- |
3, 4 |
303 |
|
AK021783.1 |
- |
1, 2 |
103 |
|
AK022770.1 |
- |
2, 3 |
202 |
|
AK024021.1 |
- |
3, 4 |
315 |
|
BC020522.1 |
- |
3, 4 |
283 |
3916886 |
CX166555.1 |
- |
4, 5 |
480 |
|
DA609590.1 |
- |
2, 3 |
441 |
|
DB091654.1 |
- |
3, 4 |
488 |
|
DB233011.1 |
- |
4, 5 |
409 |
|
View Full Product Details